missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Alpha-L-iduronidase Protein
A DNA sequence encoding the Homo sapiens (Human) Alpha-L-iduronidase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6197-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000194.2 | |
Alpha-L-iduronidase Protein | |
Research Use Only | |
IDUA | |
Recombinant Protein | |
Yeast | |
APHLVHVDAA RALWPLRRFW RSTGFCPPLP HSQADQYVLS WDQQLNLAYV GAVPHRGIKQ VRTHWLLELV TTRGSTGRGL SYNFTHLDGY LDLLRENQLL PGFELMGSAS GHFTDFEDKQ QVFEWKDLVS SLARRYIGRY GLAHVSKWNF ETWNEPDHHD FDNVSMTMQG FLNYYDACSE GLRAASPALR LGGPGDSFHT PPRSPLSWGL LRHCHDGTNF FTGEAGVRLD YISLHRKGAR SSISILEQEK VVAQQIRQLF PKFADTPIYN DEADPLVGWS LPQPWRADVT YAAMVVKVIA QHQNLLLANT TSAFPYALLS NDNAFLSYHP HPFAQRTLTA RFQVNNTRPP HVQLLRKPVL TAMGLLALLD EEQLWAEVSQ AGTVLDSNHT VGVLASAHRP QGPADAWRAA VLIYASDDTR AHPNRSVAVT LRLRGVPPGP GLVYVTRYLD NGLCSPDGEW RRLGRPVFPT AEQFRRMRAA EDPVAAAPRP LPAGGRLTLR PALRLPSLLL VHVCARPEKP PGQVTRLRAL PLTQGQLVLV WSDEHVGSKC LWTYEIQFSQ DGKAYTPVSR KPSTFNLFVF SPDTGAVSGS YRVRALDYWA RPGPFSDPVP YLEVPVPRGP PSPGNP | |
Greater than 90% as determined by SDS-PAGE. |
3425 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |