missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human alpha-Galactosidase A / GLA Protein

A DNA sequence encoding the Homo sapiens (Human) alpha-Galactosidase A / GLA, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6086-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15909869

  • 1210.00€ / 500 microgramos


Fecha estimada de envío
20-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_000160.1
alpha-Galactosidase A / GLA Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
2717
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GLA
Recombinant Protein
E. coli
LDNGLARTPT MGWLHWERFM CNLDCQEEPD SCISEKLFME MAELMVSEGW KDAGYEYLCI DDCWMAPQRD SEGRLQADPQ RFPHGIRQLA NYVHSKGLKL GIYADVGNKT CAGFPGSFGY YDIDAQTFAD WGVDLLKFDG CYCDSLENLA DGYKHMSLAL NRTGRSIVYS CEWPLYMWPF QKPNYTEIRQ YCNHWRNFAD IDDSWKSIKS ILDWTSFNQE RIVDVAGPGG WNDPDMLVIG NFGLSWNQQV TQMALWAIMA APLFMSNDLR HISPQAKALL QDKDVIAINQ DPLGKQGYQL RQGDNFEVWE RPLSGLAWAV AMINRQEIGG PRSYTIAVAS LGKGVACNPA CFITQLLPVK RKLGFYEWTS RLRSHINPTG TVLLQLENTM QMSLKDLL
Videos
Documentos