missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ALDH5A1, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) ALDH5A1, mitochondrial, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5645-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916689

  • 253.00€ / 50 microgramos


Fecha estimada de envío
22-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001071.1
ALDH5A1, mitochondrial Protein
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
7915
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ALDH5A1
Recombinant Protein
E. coli
AGRLAGLSAA LLRTDSFVGG RWLPAAATFP VQDPASGAAL GMVADCGVRE ARAAVRAAYE AFCRWREVSA KERSSLLRKW YNLMIQNKDD LARIITAESG KPLKEAHGEI LYSAFFLEWF SEEARRVYGD IIHTPAKDRR ALVLKQPIGV AAVITPWNFP SAMITRKVGA ALAAGCTVVV KPAEDTPFSA LALAELASQA GIPSGVYNVI PCSRKNAKEV GEAICTDPLV SKISFTGSTT TGKILLHHAA NSVKRVSMEL GGLAPFIVFD SANVDMS-QA VAGAMASKFR NTGQTCVCSN QFLVQRGIHD AFVKAFAEAM KKNLRVGNGF EEGTTQGPLI NEKAVEKVEK QVNDAVSKGA TVVTGGKRHQ LGKNFFEPTL LCNVTQDMLC THEETFGPLA PVIKFDTEEE AIAIANAADV GLAGYFYSQD PAQIWRVAEQ LEVGMVGVNE GLISSVECPF GGVKQSGLGR EGSKYGIDEY LELKYVCYGG L
Videos