missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Alcohol dehydrogenase class-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Alcohol dehydrogenase class-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5616-ye-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000662.3 | |
Alcohol dehydrogenase class-3 Protein | |
Research Use Only | |
ADH5 | |
Recombinant Protein | |
Yeast | |
ANEVIKCKAA VAWEAGKPLS IEEIEVAPPK AHEVRIKIIA TAVCHTDAYT LSGADPEGCF PVILGHEGAG IVESVGEGVT KLKAGDTVIP LYIPQCGECK FCLNPKTNLC QKIRVTQGKG LMPDGTSRFT CKGKTILHYM GTSTFSEYTV VADISVAKID PLAPLDKVCL LGCGISTGYG AAVNTAKLEP GSVCAVFGLG GVGLAVIMGC KVAGASRIIG VDINKDKFAR AKEFGATECI NPQDFSKPIQ EVLIEMTDGG VDYSFECIGN VKVMRAALEA CHKGWGVSVV VGVAASGEEI ATRPFQLVTG RTWKGTAFGG WKSVESVPKL VSEYMSKKIK VDEFVTHNLS FDEINKAFEL MHSGKSIRTV VKI | |
Greater than 90% as determined by SDS-PAGE. |
128 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |