missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ALCAM (CD166) Protein
A cDNA sequence encoding the ALCAM (CD166) was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10998-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
ALCAM (CD166) Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPWYTVNSA YGDTIIIPCR LDVPQNLMFG KWKYEKPDGS PVFIAFRSST KKSVQYDDVP EYKDRLNLSE NYTLSISNAR ISDEKRFVCM LVTEDNVFEA PTIVKVFKQP SKPEIVSKAL FLETEQLKKL GDCISEDSYP DGNITWYRNG KVLHPLEGAV VIIFKKEMDP VTQLYTMTSTLEYKTTKADI QMPFTCSVTY YGPSGQKTIH SEQAVFDIYY PTEQVTIQVL PPKNAIKEGD NITLKCLGNG NPPPEEFLFY LPGQPEGIRS SNTYTLTDVR RNATGDYKCS LIDKKSMIAS TAITVHYLDL SLNPSGEVTR QIGDALPVSC TISASRNATV VWMKDNIRLR SSPSFSSLHYQDAGNYVCET ALQEVEGLKK RESLTLIVEG KPQIKMTKKT DPSGLSKTII CHVEGFPKPA IQWTITGSGS VINQTEESPY INGRYYSKII ISPEENVTLT CTAENQLERT VNSLNVSANE NREKVNDQAK LIVGIVVGLL LAALEPKSCD KTHTCPPCPA PELLGGPSVF LFPPKPKDTLMISRTPEVTC VVVDVSHEDP EVKFNWYVDG VEVHNAKTKP REEQYNSTYR VVSVLTVLHQ DWLNGKEYKC KVSNKALPAP IEKTISKAKG QPREPQVYTL PPSRDELTKN QVSLTCLVKG FYPSDIAVEW ESNGQPENNY KTTPPVLDSD GSFFLYSKLT VDKSRWQQGN VFSCSVMHEALHNHYTQKSL SLSPGKHHHH HH | |
Greater than 90.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CD166 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |