missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AKT-interacting Protein

A DNA sequence encoding the Homo sapiens (Human) AKT-interacting protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8053-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052391

  • 234.00€ / 50 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001012398.1
AKT-interacting Protein
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
64400
Greater than 90% as determined by SDS-PAGE.
Research Use Only
AKTIP
Recombinant Protein
E. coli
MNPFWSMSTS SVRKRSEGEE KTLTGDVKTS PPRTAPKKQL PSIPKNALPI TKPTSPAPAA QSTNGTHASY GPFYLEYSLL AEFTLVVKQK LPGVYVQPSY RSALMWFGVI FIRHGLYQDG VFKFTVYIPD NYPDGDCPRL VFDIPVFHPL VDPTSGELDV KRAFAKWRRN HNHIWQVLMY ARRVFYKIDT ASPLNPEAAV LYEKDIQLFK SKVVDSVKVC TARLFDQPKI EDPYAISFSP WNPSVHDEAR EKMLTQKKPE EQHNKSVHVA GLSWVKPGSV QPFSKEEKTV AT
Videos
Documentos