missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AK3L1 Protein
A cDNA sequence encoding the AK3L1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10990-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
205 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMASK LLRAVILGPP GSGKGTVSQR IAQNFGLQHL SSGHFLRENIKASTEVGEMA KQYIEKSLLV PDHVITRLMM SELENRRGQH WLLDGFPRTL GQAEALDKIC EVDLVISLNI PFETLKDRLS RRWIHPPSGR VYNLDFNPPH VHGIDDVTGE PLVQQEDDKP EAVAARLRQY KDVAKPVIEL YKSRGVLHQF SGTETNKIWP YVYTLFSNKI TPIQSKEAY | |
Greater than 90.0% as determined by SDS-PAGE. |
AK3L1 Protein | |
Research Use Only | |
AK4 | |
Human | |
His | |
AK3L1 solution containing 20mM Tris-HCl pH 8.0, 2mM DTT and 20% glycerol. |