missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AFP Protein
A cDNA sequence encoding the AFP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10975-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
AFP Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
MKWVESIFLI FLLNFTESRT LHRNEYGIAS ILDSYQCTAE ISLADLATIF FAQFVQEATY KEVSKMVKDA LTAIEKPTGD EQSSGCLENQ LPAFLEELCH EKEILEKYGH SDCCSQSEEG RHNCFLAHKK PTPASIPLFQ VPEPVTSCEA YEEDRETFMN KFIYEIARRH PFLYAPTILL WAARYDKIIP SCCKAENAVE CFQTKAATVT KELRESSLLN QHACAVMKNF GTRTFQAITV TKLSQKFTKV NFTEIQKLVL DVAHVHEHCC RGDVLDCLQD GEKIMSYICS QQDTLSNKIT ECCKLTTLER GQCIIHAEND EKPEGLSPNL NRFLGDRDFN QFSSGEKNIF LASFVHEYSR RHPQLAVSVI LRVAKGYQEL LEKCFQTENP LECQDKGEEE LQKYIQESQA LAKRSCGLFQ KLGEYYLQNA FLVAYTKKAP QLTSSELMAI TRKMAATAAT CCQLSEDKLL ACGEGAADII IGHLCIRHEM TPVNPGVGQC CTSSYANRRP CFSSLVVDET YVPPAFSDDK FIFHKDLCQA QGVALQTMKQ EFLINLVKQK PQITEEQLEA VIADFSGLLE KCCQGQEQEV CFAEEGQKLI SKTRAALGV |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Alpha Fetoprotein protein at 2.5 mg/ml 0.1M PBS buffer pH 7.4 containing 0.1% NaN3. |