missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADPRHL2 Protein

A cDNA sequence encoding the ADPRHL2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10972-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915738

  • 3091.00€ / 1 miligramo


Fecha estimada de envío
17-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

54936
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ADPRHL2
Human
His
The ADPRHL2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 30% glycerol and 1mM DTT.
ADPRHL2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAAAAM AAAAGGGAGA ARSLSRFRGC LAGALLGDCV GSFYEAHDTV DLTSVLRHVQ SLEPDPGTPG SERTEALYYT DDTAMARALV QSLLAKEAFD EVDMAHRFAQ EYKKDPDRGY GAGVVTVFKK LLNPKCRDVF EPARAQFNGK GSYGNGGAMR VAGISLAYSS VQDVQKFARL SAQLTHASSL GYNGAILQAL AVHLALQGES SSEHFLKQLL GHMEDLEGDA QSVLDARELG MEERPYSSRL KKIGELLDQA SVTREEVVSE LGNGIAAFES VPTAIYCFLR CMEPDPEIPS AFNSLQRTLI YSISLGGDTD TIATMAGAIA GAYYGMDQVP ESWQQSCEGY EETDILAQSL HRVFQKS
Videos