missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADP-ribosylation factor-like protein 4A Protein
A DNA sequence encoding the Homo sapiens (Human) ADP-ribosylation factor-like protein 4A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5678-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001032241.1 | |
ADP-ribosylation factor-like protein 4A Protein | |
Research Use Only | |
ARL4A | |
Recombinant Protein | |
E. coli | |
GNGLSDQTSI LSNLPSFQSF HIVILGLDCA GKTTVLYRLQ FNEFVNTVPT KGFNTEKIKV TLGNSKTVTF HFWDVGGQEK LRPLWKSYTR CTDGIVFVVD SVDVERMEEA KTELHKITRI SENQGVPVLI VANKQDLRNS LSLSEIEKLL AMGELSSSTP WHLQPTCAII GDGLKEGLEK LHDMIIKRRK MLRQQKKKR | |
Greater than 90% as determined by SDS-PAGE. |
10124 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |