missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADP-ribosylation factor-like protein 1

A DNA sequence encoding the Homo sapiens (Human) ADP-ribosylation factor-like protein 6, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8213-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053321

  • 234.00€ / 50 microgramos


Fecha estimada de envío
19-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001265222.1
ADP-ribosylation factor-like protein 1
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
84100
Greater than 80% as determined by SDS-PAGE.
Research Use Only
ARL6
Recombinant Protein
E. coli
GLLDRLSVLL GLKKKEVHVL CLGLDNSGKT TIINKLKPSN AQSQNILPTI GFSIEKFKSS SLSFTVFDMS GQGRYRNLWE HYYKEGQAII FVIDSSDRLR MVVAKEELDT LLNHPDIKHR RIPILFFANK MDLRDAVTSV KVSQLLCLEN IKDKPWHICA SDAIKGEGLQ EGVDWLQDQI QTVKT
Videos
Documentos