missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADK Protein

A cDNA sequence encoding the ADK was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10507-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914088

  • 103.00€ / 2 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

132
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
ADK
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MRENILFGMG NPLLDISAVV DKDFLDKYSL KPNDQILAED KHKELFDELV KKFKVEYHAG GSTQNSIKVA QWMIQQPHKA ATFFGCIGID KFGEILKRKA AEAHVDAHYY EQNEQPTGTC AACITGDNRS LIANLAAANC YKKEKHLDLE KNWMLVEKARVCYIAGFFLT VSPESVLKVA HHASENNRIF TLNLSAPFIS QFYKESLMKV MPYVDILFGN ETEAATFARE QGFETKDIKE IAKKTQALPK MNSKRQRIVI FTQGRDDTIM ATESEVTAFA VLDQDQKEII DTNGAGDAFV GGFLSQLVSD KPLTECIRAG HYAASIIIRR TGCTFPEKPDFH
ADK Protein
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 30 pmol/min/ug and is defined as the amount of enzyme that convert 1.0 pmole of adenosine to AMP per minute at pH 7.5 at 37C in a couple system with PK and LDH.
Human
His
The ADK protein solution (0.5 mg/ml) contains 20% glycerol, 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 1mM EDTA and 50mM NaCl.
Videos