missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADI1 Protein
A cDNA sequence encoding the ADI1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10968-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
ADI1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSSMVL AWYMDDAPGD PRQPHRPDPG RPVGLEQLRR LGVLYWKLDA DKYENDPELE KIRRERNYSW MDIITICKDK LPNYEEKIKM FYEEHLHLDD EIRYILDGSG YFDVRDKEDQ WIRIFMEKGD MVTLPAGIYH RFTVDEKNYT KAMRLFVGEP VWTAYNRPAD HFEARGQYVK FLAQTA | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ADI1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |