missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADH7 Protein

A DNA sequence encoding the Homo sapiens (Human) ADH7, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5617-ye-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916479

  • 3391.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

NP_000664.2
ADH7 Protein
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
131
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ADH7
Recombinant Protein
Yeast
MFAEIQIQDK DRMGTAGKVI KCKAAVLWEQ KQPFSIEEIE VAPPKTKEVR IKILATGICR TDDHVIKGTM VSKFPVIVGH EATGIVESIG EGVTTVKPGD KVIPLFLPQC RECNACRNPD GNLCIRSDIT GRGVLADGTT RFTCKGKPVH HFMNTSTFTE YTVVDESSVA KIDDAAPPEK VCLIGCGFST GYGAAVKTGK VKPGSTCVVF GLGGVGLSVI MGCKSAGASR IIGIDLNKDK FEKAMAVGAT ECISPKDSTK PISEVLSEMT GNNVGYTFEV IGHLETMIDA LASCHMNYGT SVVVGVPPSA KMLTYDPMLL FTGRTWKGCV FGGLKSRDDV PKLVTEFLAK KFDLDQLITH VLPFKKISEG FELLNSGQSI RTVLTF
Videos