missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Activin A / INHBA Protein

A DNA sequence encoding the Homo sapiens (Human) Activin A / INHBA Protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6232-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16051790

  • 1180.00€ / 500 microgramos


Fecha estimada de envío
20-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_002183.1
Activin A / INHBA Protein
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
3624
Greater than 90% as determined by SDS-PAGE.
Research Use Only
INHBA
Recombinant Protein
E. coli
GLECDGKVNI CCKKQFFVSF KDIGWNDWII APSGYHANYC EGECPSHIAG TSGSSLSFHS TVINHYRMRG HSPFANLKSC CVPTKLRPMS MLYYDDGQNI IKKDIQNMIV EECGCS
Videos
Documentos