missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Actin, cytoplasmic 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Actin, cytoplasmic 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8379-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054311

  • 234.00€ / 50 microgramos


Fecha estimada de envío
21-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001092.1
Actin, cytoplasmic 1 Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
60
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ACTB
Recombinant Protein
E. coli
PVLLTEAPLN PKANREKMTQ IMFETFNTPA MYVAIQAVLS LYASGRTTGI VMDSGDGVTH TVPIYEGYAL PHAILRLDLA GRDLTDYLMK ILTERGYSFT TTAEREIVRD IKEKLCYVAL DFEQEMATAA SSSSLEKSYE LPDGQVITIG NERFRCPEAL FQPSFLGMES CGIHETTFNS IMKCDVDIRK DLYANTVLSG GTTMYPGIAD RMQKEITALA PSTMKIKIIA PPERKYSVWI GGSILASLST FQQMWISKQE YDESGPSIVH RKCF
Videos
Documentos