missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACP6 Protein

A cDNA sequence encoding the ACP6 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10503-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15994068

Cantidad Precio
1 129.74€ / 5 microgramos
10 121.33€ / 5 microgramos
Envío estimado: 04-03-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.



Descripción

Descripción

Especificaciones

Especificaciones

ACP6 Protein
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSELQEADG QCPVDRSLLK LKMVQVVFRH GARSPLKPLP LEEQVEWNPQ LLEVPPQTQF DYTVTNLAGG PKPYSPYDSQ YHETTLKGGM FAGQLTKVGM QQMFALGERL RKNYVEDIPF LSPTFNPQEV FIRSTNIFRN LESTRCLLAG LFQCQKEGPI IIHTDEADSE VLYPNYQSCW SLRQRTRGRR QTASLQPGIS EDLKKVKDRM GIDSSDKVDF FILLDNVAAE QAHNLPSCPM LKRFARMIEQ RAVDTSLYIL PKEDRESLQM AVGPFLHILE SNLLKAMDSA TAPDKIRKLY LYAAHDVTFI PLLMTLGIFD HKWPPFAVDL TMELYQHLES KEWFVQLYYH GKEQVPRGCP DGLCPLDMFL NAMSVYTLSP EKYHALCSQT QVMEVGNEE
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Specific activity is > 70 units/mg, and is defined as the amount of enzyme which hydrolyze 1.0 nmoles of p-nitrophenyl phosphate (pNPP) per minute at PH 5.0 at 37C.
Human
His
ACP6 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 30% glycerol and 1mM DTT.
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales