missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACOT8 Protein

A cDNA sequence encoding the ACOT8 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10944-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15935648

  • 161.00€ / 20 microgramos


Fecha estimada de envío
29-06-2020
Add to basket

Specifications

Specifications

10005
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
ACOT8
Human
His
The ACOT8 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol and 2mM DTT.
ACOT8 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSSPQAP EDGQGCGDRG DPPGDLRSVL VTTVLNLEPL DEDLFRGRHY WVPAKRLFGG QIVGQALVAA AKSVSEDVHV HSLHCYFVRA GDPKLPVLYQ VERTRTGSSF SVRSVKAVQH GKPIFICQAS FQQAQPSPMQ HQFSMPTVPP PEELLDCETL IDQYLRDPNL QKRYPLALNR IAAQEVPIEI KPVNPSPLSQ LQRMEPKQMF WVRARGYIGE GDMKMHCCVA AYISDYAFLG TALLPHQWQH KVHFMVSLDH SMWFHAPFRA DHWMLYECES PWAGGSRGLV HGRLWRQDGV LAVTCAQEGV IRVKPQVSES KL
Videos
Documents