missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACOT7 Protein
A cDNA sequence encoding the ACOT7 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10943-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
11332 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MARPGLIHSA PGLPDTCALL QPPAASAAAA PSMSGPDVET PSAIQICRIM RPDDANVAGN VHGGTILKMI EEAGAIISTR HCNSQNGERC VAALARVERT DFLSPMCIGE VAHVSAEITY TSKHSVEVQV NVMSENILTG AKKLTNKATL WYVPLSLKNV DKVLEVPPVV YSRQEQEEEG RKRYEAQKLE RMETKWRNGD IVQPVLNPEP NTVSYSQSSL IHLVGPSDCT LHGFVHGGVT MKLMDEVAGI VAARHCKTNI VTASVDAINF HDKIRKGCVI TISGRMTFTS NKSMEIEVLV DADPVVDSSQ KRYRAASAFF TYVSLSQEGR SLPVPQLVPE TEDEKKRFEE GKGRYLQMKA KRQGHAEPQP | |
Greater than 95.0% as determined by SDS-PAGE. |
ACOT7 Protein | |
Research Use Only | |
ACOT7 | |
Human | |
His | |
The ACOT7 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.15M NaCl. |