missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACLY / acly / ATP citrate lyase Protein

A DNA sequence encoding the Homo sapiens (Human) ACLY / acly / ATP citrate lyase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5607-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916409

  • 1776.00€ / 500 microgramos


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001087.2
ACLY / acly / ATP citrate lyase Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
47
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ACLY
Recombinant Protein
E. coli
KAISEQTGKE LLYKFICTTS AIQNRFKYAR VTPDTDWARL LQDHPWLLSQ NLVVKPDQLI KRRGKLGLVG VNLTLDGVKS WLKPRLGQEA TVGKATGFLK NFLIEPFVPH SQAEEFYVCI YATREGDYVL FHHEGGVDVG DVDAKAQKLL VGVDEKLNPE DIKKHLLVHA PEDKKEILAS FISGLFNFYE DLYFTYLEIN PLVVTKDGVY VLDLAAKVDA TADYICKVKW GDIEFPPPFG REAYPEEAYI ADLDAKSGAS LK
Videos