missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Acid ceramidase Protein

A DNA sequence encoding the Homo sapiens (Human) Acid ceramidase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7569-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16049460

  • 1180.00€ / 500 microgramos


Fecha estimada de envío
19-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001120977.1
Acid ceramidase Protein
500 μg
Not determined.
Full Length of Mature Protein
Human
His-SUMO
Tris-based buffer, 50% glycerol
427
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ASAH1
Recombinant Protein
E. coli
QHAPPWTEDC RKSTYPPSGP TYRGAVPWYT INLDLPPYKR WHELMLDKAP VLKVIVNSLK NMINTFVPSG KIMQVVDEKL PGLLGNFPGP FEEEMKGIAA VTDIPLGEII SFNIFYELFT ICTSIVAEDK KGHLIHGRNM DFGVFLGWNI NNDTWVITEQ LKPLTVNLDF QRNNKTVFKA SSFAGYVGML TGFKPGLFSL TLNERFSING GYLGILEWIL GKKDVMWIGF LTRTVLENST SYEEAKNLLT KTKILAPAYF ILGGNQSGEG CVITRDRKES LDVYELDAKQ GRWYVVQTNY DRWKHPFFLD DRRTPAKMCL NRTSQENISF ETMYDVLSTK PVLNKLTVYT TLIDVTKGQF ETYLRDCPDP CIGW
Videos
Documentos