missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Acetylcholine receptor subunit alpha Protein

A DNA sequence encoding the Homo sapiens (Human) Acetylcholine receptor subunit alpha, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5832-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15908099

  • 183.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

NP_000070.1
Acetylcholine receptor subunit alpha Protein
10 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
1134
Greater than 82.8% as determined by SDS-PAGE.
Research Use Only
CHRNA1
Recombinant Protein
E. coli
SEHETRLVAK LFKDYSSVVR PVEDHRQVVE VTVGLQLIQL INVDEVNQIV TTNVRLKQGD MVDLPRPSCV TLGVPLFSHL QNEQWVDYNL KWNPDDYGGV KKIHIPSEKI WRPDLVLYNN ADGDFAIVKF TKVLLQYTGH ITWTPPAIFK SYCEIIVTHF PFDEQNCSMK LGTWTYDGSV VAINPESDQP DLSNFMESGE WVIKESRGWK HSVTYSCCPD TPYLDITYHF VMQRL
Videos