missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ABCB8 Protein
A DNA sequence encoding the Homo sapiens(Human) ABCB8, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10021-iv-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001269220.1 | |
ABCB8 Protein | |
Research Use Only | |
ABCB8 | |
Recombinant Protein | |
E. coli | |
GKTGQLHKAE GEYSDGYRSS SLLRAVAHLR SQLWAHLPRA PLAPRWSPSA WCWVGGALLG PMVLSKHPHL CLVALCEAEE APPASSTPHV VGSRFNWKLF WQFLHPHLLV LGVAVVLALG AALVNVQIPL LLGQLVEVVA KYTRDHVGSF MTESQNLSTH LLILYGVQGL LTFGYLVLLS HVGERMAVDM RRALFSSLLR QDITFFDANK TGQLVSRLTT DVQEFKSSFK LVISQGLRSC TQVAGCLVSL SMLSTRLTLL LMVATPALMG VGTLMGSGLR KLSRQCQEQI ARAMGVADEA LGNVRTVRAF AMEQREEERY GAELEACRCR AEELGRGIAL FQGLSNIAFN CMVLGTLFIG GSLVAGQQLT GGDLMSFLVA SQTVQRSMAN LSVLFGQVVR GLSAGARVFE YMALNPCIPL SGGCCVPKEQ LRGSVTFQNV CFSYPCRPGF EVLKDFTLTL PPGKIVALVG QSGGGKTTVA SLLERFYDPT AGVVMLDGRD LRTLDPSWLR GQVVGFISQE PVLFGTTIME NIRFGKLEAS DEEVYTAARE ANAHEFITSF PEGYNTVVGE RGTTLSGGQK QRLAIARALI KQPTVLILDE ATSALDAESE RVVQEALDRA SAGRTVLVIA HRLSTVRGAH CIVVMADGRV WEAGTH | |
Greater than 85% as determined by SDS-PAGE. |
11194 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |