missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L5 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8412-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000960.2 | |
60S ribosomal protein L5 Protein | |
Research Use Only | |
RPL5 | |
Recombinant Protein | |
E. coli | |
GFVKVVKNKA YFKRYQVKFR RRREGKTDYY ARKRLVIQDK NKYNTPKYRM IVRVTNRDII CQIAYARIEG DMIVCAAYAH ELPKYGVKVG LTNYAAAYCT GLLLARRLLN RFGMDKIYEG QVEVTGDEYN VESIDGQPGA FTCYLDAGLA RTTTGNKVFG ALKGAVDGGL SIPHSTKRFP GYDSESKEFN AEVHRKHIMG QNVADYMRYL MEEDEDAYKK QFSQYIKNSV TPDMMEEMYK KAHAAIRENP VYEKKPKKEV KKKRWNRPKM SLAQKKDRVA QKKASFLRAQ ERAAES | |
Greater than 90% as determined by SDS-PAGE. |
6125 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |