missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 60S ribosomal protein L10a Protein

A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L10a, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8500-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054971

  • 253.00€ / 50 microgramos


Fecha estimada de envío
26-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_009035.3
60S ribosomal protein L10a Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
4736
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RPL10A
Recombinant Protein
E. coli
KVSRDTLYEA VREVLHGNQR KRRKFLETVE LQISLKNYDP QKDKRFSGTV RLKSTPRPKF SVCVLGDQQH CDEAKAVDIP HMDIEALKKL NKNKKLVKKL AKKYDAFLAS ESLIKQIPRI LGPGLNKAGK FPSLLTHNEN MVAKVDEVKS TIKFQMKKVL CLAVAVGHVK MTDDELVYNI HLAVNFLVSL LKKNWQNVRA LYIKSTMGKP QR
Videos
Documentos