missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 60S acidic ribosomal protein P2 Protein

A DNA sequence encoding the Homo sapiens (Human) 60S acidic ribosomal protein P2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8404-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054421

  • 160.00€ / 10 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000995.1
60S acidic ribosomal protein P2 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
6181
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RPLP2
Recombinant Protein
E. coli
MRYVASYLLA ALGGNSSPSA KDIKKILDSV GIEADDDRLN KVISELNGKN IEDVIAQGIG KLASVPAGGA VAVSAAPGSA APAAGSAPAA AEEKKDEKKE ESEESDDDMG FGL
Videos
Documentos