missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S acidic ribosomal protein P2 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S acidic ribosomal protein P2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8404-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000995.1 | |
60S acidic ribosomal protein P2 Protein | |
Research Use Only | |
RPLP2 | |
Recombinant Protein | |
E. coli | |
MRYVASYLLA ALGGNSSPSA KDIKKILDSV GIEADDDRLN KVISELNGKN IEDVIAQGIG KLASVPAGGA VAVSAAPGSA APAAGSAPAA AEEKKDEKKE ESEESDDDMG FGL | |
Greater than 90% as determined by SDS-PAGE. |
6181 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |