missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 43358 Protein
A cDNA sequence encoding the 43358 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10916-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
43358 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSVSAFGAE FSSEACRELG FSSNLLCSSC DLLGQFNLLQ LDPDCRGCCQ EEAQFETKKL YAGAILEVCG CKLGRFPQVQ AFVRSDKPKL FRGLQIKYVR GSDPVLKLLD DNGNIAEELS ILKWNTDSVE EFLSEKLERI | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SEP15 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |