missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 39S ribosomal protein L20, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L20, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8062-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_060441.2 | |
39S ribosomal protein L20, mitochondrial Protein | |
Research Use Only | |
MRPL20 | |
Recombinant Protein | |
E. coli | |
VIRAFVKCTK ARYLKKKNMR TLWINRITAA SQEHGLKYPA LIGNLVKCQV ELNRKVLADL AIYEPKTFKS LAALASRRRH EGFAAALGDG KEPEGIFSRV VQYH | |
Greater than 90% as determined by SDS-PAGE. |
55052 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |