missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 10 kDa heat shock protein, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) 10 kDa heat shock protein, mitochondrial, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP1136-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15917018

  • 1210.00€ / 500 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_002148.1
10 kDa heat shock protein, mitochondrial Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
3336
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HSPE1
Recombinant Protein
E. coli
AGQAFRKFLP LFDRVLVERS AAETVTKGGI MLPEKSQGKV LQATVVAVGS GSKGKGGEIQ PVSVKVGDKV LLPEYGGTKV VLDDKDYFLF RDGDILGKYV D
Videos