missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HCV NS5B Protein
A cDNA sequence encoding the HCV NS5B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12201-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HSV HCV NS5B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSPMGF SYDTRCFDST VTESDIRTEE AIYQCCDLDP QARVAIKSLT ERLYVGGPLT NSRGENCGYR RCRASGVLTT SCGNTLTCYI KARAACRAAG LQDCTMLVCG DDLVVICESA GVQED | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
His | |
The HCV NS5B protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |