missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Horse Erythropoietin / EPO Protein

A DNA sequence encoding the Equus caballus (Horse) Erythropoietin / EPO, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5988-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919179

  • 472.00€ / 50 microgramos


Fecha de envío estimada
del proveedor 04-04-2019
Añadir a la cesta

Especificación

Especificación

NP_001075294.1
Erythropoietin / EPO Protein
50 μg
Not determined.
Full Length
Horse
His-SUMO
Tris-based buffer, 50% glycerol
100033849
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EPO
Recombinant Protein
E. coli
APPRLICDSR VLERYILEAR EAENVTMGCA EGCSFGENVT VPDTKVNFYS WKRMEVEQQA VEVWQGLALL SEAILQGQAL LANSSQPSET LRLHVDKAVS SLRSLTSLLR ALGAQKEAIS PPDAASAAPL RTFAVDTLCK LFRIYSNFLR GKLKLYTGEA CRRGDR
Videos