missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-2 gp32 Protein

A cDNA sequence encoding the HIV-2 gp32 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12268-EC-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919888

  • 691.00€ / 500 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

HIV-2 gp32 Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
The HIV-2 gp32 solution (1 mg/ml) 0.01M Na2CO3, 0.01M Na3EDTA, 0.014 Mb-mercaptoethanol and 0.02% Sarcosyl.
Videos