missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 p55 gag Protein
A cDNA sequence encoding the HIV-1 p55 gag was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12258-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HIV-1 p55 gag Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The HIV-1 p55 gag protein solution contains 10mM Tris pH 8.0, 140mM NaCl and 400mM L-Arginine. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 90.0% as determined by HPLC analysis & SDS-PAGE. |