missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 gp41 Protein
A cDNA sequence encoding the HIV-1 gp41 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12248-EC-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HIV-1 gp41 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
8M urea, 20mM Tris-HCl pH 8.0, 10mM B-mercaptoethanol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |