missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant hepatitis Capsid Protein

A DNA sequence encoding the Hepatitis E virus genotype 3 (isolate Human/United States/US2) (HEV-3) Capsid protein, was expressed in the hosts and tags indicated. option, abov

Marca:  enQuireBio™ QP8294-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053831

  • 1557.00€ / 1 miligramo


Fecha estimada de envío
23-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

hepatitis Capsid Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Hepatitis
His
Tris-based buffer, 50% glycerol
Greater than 80% as determined by SDS-PAGE.
Research Use Only
ORF2
Recombinant Protein
E. coli
KLYTSVENAQ QDKGITIPHD IDLGDSRVVI QDYDNQHEQD RPTPSPAPSR PFSVLRANDV LWLSLTAAEY DQTTYGSSTN PMYVSDTVTL VNVATGAMS- QAVARSLDWS KVTLDGRPLT TIQQYSKTFY VLPLRGKLSF WEAGTTKAGY PYNYNTTASD QILIENAAGH RVAISTYTTS LGAGPTSISA VGVLAPHSAL AVLEDTIDYP ARAHTFDDFC PECRTLGLQG CAFQSTIAEL QRLKMKVGKT RES
Videos
Documentos