missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS4 Mosaic Protein
A cDNA sequence encoding the HCV NS4 Mosaic was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12180-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Hepatitis C HCV NS4 Mosaic Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris, pH 8.0, 5mM EDTA. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
His | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |