missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV NS4 Mosaic Protein

A cDNA sequence encoding the HCV NS4 Mosaic was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12180-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919618

  • 691.00€ / 500 microgramos


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

Hepatitis C HCV NS4 Mosaic Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
His
Protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
50mM Tris, pH 8.0, 5mM EDTA.
Videos