missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV NS3 Genotype-6 Protein

A cDNA sequence encoding the HCV NS3 Genotype-6 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12173-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919598

  • 183.00€ / 100 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

Hepatitis C HCV NS3 Genotype-6 Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
GST
HCV NS3 Genotype-6 protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X and 50% Glycerol.
Videos