missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS3 Genotype-1c Protein
A cDNA sequence encoding the HCV NS3 Genotype-1c was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12166-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Hepatitis C HCV NS3 Genotype-1c Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X and 50% Glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
His | |
HCV NS3 Genotype-1c protein is >95% pure as determined by 10% PAGE (coomassie staining). |