missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS3 Genotype-1b Protein
A cDNA sequence encoding the HCV NS3 Genotype-1b (Amino Acids 1356-1459) was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10428-ec-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005441.1 | |
Hepatitis C HCV NS3 Genotype-1b Protein | |
Research Use Only | |
NOG | |
The ED50 as determined by inhibiting BMP-4- induced alkaline phosphatase production of murine ATDC5 cells is less than 3.0 ng/ml, corresponding to a specific activity of > 3.3 M 105 IU/mg in the presence of 5 ng/ml rHuBMP-4. | |
Human | |
Untagged | |
0.2 um filtered 30 % acetonitrile, 0.1 % TFA, lyophilized |
9241 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MQHYLHIRPA PSDNLPLVDL IEHPDPIFDP KEKDLNETLL RSLLGGHYDP GFMATSPPED RPGGGGGAAG GAEDLAELDQ LLRQRPSGAM PSEIKGLEFS EGLAQGKKQR LSKKLRRKLQ MWLWSQTFCP VLYAWNDLGS RFWPRYVKVG SCFSKRSCSV PEGMVCKPSK SVHLTVLRWR CQRRGGQRCG WIPIQYPIIS ECKCSC | |
Greater than 95% as determined by SDS-PAGE and HPLC. |