missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV NS3 Genotype-1b Protein

A cDNA sequence encoding the HCV NS3 Genotype-1b (Amino Acids 1356-1459) was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10428-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913828

  • 1510.00€ / 250 microgramos


Fecha estimada de envío
23-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_005441.1
Hepatitis C HCV NS3 Genotype-1b Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MQHYLHIRPA PSDNLPLVDL IEHPDPIFDP KEKDLNETLL RSLLGGHYDP GFMATSPPED RPGGGGGAAG GAEDLAELDQ LLRQRPSGAM PSEIKGLEFS EGLAQGKKQR LSKKLRRKLQ MWLWSQTFCP VLYAWNDLGS RFWPRYVKVG SCFSKRSCSV PEGMVCKPSK SVHLTVLRWR CQRRGGQRCG WIPIQYPIIS ECKCSC
9241
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
NOG
The ED50 as determined by inhibiting BMP-4- induced alkaline phosphatase production of murine ATDC5 cells is less than 3.0 ng/ml, corresponding to a specific activity of > 3.3 M 105 IU/mg in the presence of 5 ng/ml rHuBMP-4.
Human
Untagged
0.2 um filtered 30 % acetonitrile, 0.1 % TFA, lyophilized
Videos