missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV Core Genotype-1b Protein

A cDNA sequence encoding the HCV Core Genotype-1b was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12146-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919518

  • 1372.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 08-04-2019
Añadir a la cesta

Especificación

Especificación

Hepatitis C HCV Core Genotype-1b Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
GST
HCV Core Genotype-1b protein is >95% pure as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
(1 mg/ml) 1.5M urea, 25mM Tris-HCl, pH 8.0, 0.2% Triton-X and 50% glycerol.
Videos