missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV 4th Generation Protein
A cDNA sequence encoding the HCV 4th Generation was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12136-EC-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Hepatitis C HCV 4th Generation Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Phosphate saline buffer with 25mM arginine. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |