missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant H. pylori cagA Protein

A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) cagA, was expressed in the hosts and tags indicated. option, abo

Marca:  enQuireBio™ QP8795-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16056511

  • 472.00€ / 50 microgramos


Fecha estimada de envío
23-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_207343.1
cagA Protein
50 μg
Not determined.
Partial (see sequence information for more details).
H. pylori
His
Tris-based buffer, 50% glycerol
898777
Greater than 90% as determined by SDS-PAGE.
Research Use Only
cagA
Recombinant Protein
E. coli
MTNETIDQTR TPDQTQSQTA FDPQQFINNL QVAFIKVDNV VASFDPDQKP IVDKNDRDNR QAFDGISQLR EEYSNKAIKN PTKKNQYFSD FIDKSNDLIN KDNLIDVESS TKSFQKFGDQ RYQIFTSWVS HQKDPSKINT RSIRNFMENI IQPPIPDDKE KAEFLKSAKQ SFAGIIIGNQ IRTDQKFMGV FDESLKERQE AEKNGGPTGG DWLDIFLSFI FNKKQSSDVK EAINQEPVPH VQPDIATTTT DIQGLPPEAR DLLDERGNFS KFTLGDMEML DVEGVADIDP NYKF
Videos
Documentos