missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Guinea Pig CD46 Protein
A DNA sequence encoding the Cavia porcellus (Guinea pig) CD46, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5789-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001166399.2 | |
CD46 Protein | |
Research Use Only | |
Cd46 | |
Recombinant Protein | |
E. coli | |
CVLPPPFEAM EPINPKPYYE IGEKVEYRCK KGYLRQPFYL MVATCEKNHS WVPITDDGCI KKQCTYLNPP PKGRVEYING TRTWGDIVHF SCVEGFYVSG IAALSCELRG DNVDWNGRVP TCEKVLCSPP PKIQNGKYTF SDVQVFEYFE AVTYSCDAVQ GPDKLSLVGN EVLYCAGHQK WSSAAPECKV VKCPLPVVKN GKQISGLGQT FFYQATVTFQ CLPGFYFNGS STVVCGSDNT WKPSIPECLK GPKPTHPTKP PVYNYPGYPN PREGIFDQEL N | |
Greater than 90% as determined by SDS-PAGE. |
100135498 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Guinea Pig | |
His | |
Tris-based buffer, 50% glycerol |