missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Gilthead Seabream GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10633-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
GH Protein | |
Research Use Only | |
Binding assays of the 125I-labeled gilthead seabream GH to dolphin fish liver microsomal fraction resulted in high specific binding characterized by a Ka of 1.93 nM and a Bmax of 540 fmol/mg microsomal fraction protein. Recombinant gilthead seabream Growth Hormone, like ovine placental lactogen, exhibited growth-stimulating activity when applied orally to Sparus aurata larvae or intraperitoneally to juvenile fish. | |
Gilthead Seabream | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.02% NaHCO3. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MDRVVLMLSV MSLGVSSQPI TDGQRLFSIA VSRVQHLHLL AQRLFSDFES SLQTEEQRQL NKIFLQDFCN SDYIISPIDK HETQRSSVLK LLSISYRLVE SWEFPSRSLS GGSAPRNQIS PKLSELKTGI HLLIRANEDG AEIFPDSSAL QLAPYGNYYQ SLGTDESLRR TYELLACFKK DMHKVETYLT VAKCRLSPEA NCTL | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |