missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Feline IL2 Protein
A cDNA sequence encoding the IL2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10727-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
IL2 Protein | |
Research Use Only | |
Measured in a cell proliferation assay using CTLL2 mouse cytotoxic T cells. The ED50 for this effect is less or equal to 0.3ng/ml. | |
Feline | |
His | |
IL2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl(pH 8.0), 0.15M NaCl and 30% glycerol. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSAPASSST KETQQQLEQL LLDLRLLLNG VNNPENPKLS RMLTFKFYVP KKATELTHLQ CLVEELKPLE EVLYLAQSKN FHLNHIKELM SNINVTVLKL KGSETRFTCN YDDETATIVE FLNKWITFCQ SIFSTLT | |
Greater than 95% as determined by SDS-PAGE. |