missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Equine IFN g Protein

A cDNA sequence encoding the IFN g was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10675-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15924618

  • 3091.00€ / 1 miligramo


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

100034181
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
IFNG
Recombinant Protein
E. coli
QAAFFKEIEN LKEYFNASNP DVGDGGPLFL DILKNWKEDS DKKIIQSQIV SFYFKLFENL KDNQVIQKSM DTIKEDLFVK FFNSSTSKLE DFQKLIQIPV NDLKVQRKAI SELIKVMNDL SPKANLRKRK RSQNPFRGRR ALQ
IFN g Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50 as determined by an anti-viral assay using human HeLa cells infected with encephalomyocarditis (EMC) virus is less than 10.0 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg.
Equine
Untagged
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4 and 5% trehalose.
Videos