missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant EBV EBNA1 Mosaic Protein

A cDNA sequence encoding the EBV EBNA1 Mosaic was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11732-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918178

  • 183.00€ / 100 microgramos


Fecha de envío estimada
del proveedor 09-04-2019
Añadir a la cesta

Especificación

Especificación

EBNA1 Mosaic Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
EBV
His
Protein is >95% pure as determined by SDS-PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
10mM PBS pH 7.6 and 10mM NaCl.
Videos